Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Protocadherin-12 Sheep anti-Mouse, Polyclonal, R&D Systems
Sheep Polyclonal Antibody
Manufacturer: R&D Systems AF7926SP
This item is not returnable.
View return policy
Description
Protocadherin-12 Polyclonal antibody AF7926SP specifically detects Protocadherin-12 in Mouse samples. It is validated for Western BlottingSpecifications
Protocadherin-12 | |
LYOPH | |
Western Blot 1 ug/mL | |
O55134 | |
PCDH12, protocadherin 12, Protocadherin12, Vascular cadherin-2, vascular endothelial cadherin 2, Vascular endothelial cadherin-2, VE-cad-2, VECAD2, VE-cadherin 2, VE-cadherin-2, VE-cadherin-2protocadherin-12 | |
Sheep | |
IgG | |
25 ug | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
Polyclonal | |
Detects mouse Protocadherin-12 in direct ELISAs and Western blots. In direct ELISAs, less than 1% cross-reactivity with recombinant human PCDH-18 is observed. | |
Mouse |
Western Blot | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
Affinity Purified | |
PCDH12 | |
Mouse myeloma cell line NS0-derived recombinant mouse Protocadherin-12 Leu18-Ala716 Accession # O55134 | |
Immunogen affinity purified | |
RUO | |
Primary | |
51294 | |
Sterile PBS to a final concentration of 0.2 mg/mL. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only