Learn More
Protocadherin-19 Mouse anti-Human, Alexa Fluor 350, Clone: 921614, R&D Systems™
Mouse MonoclonalAntibody
Manufacturer:  R&D Systems FAB8626U100UG
Catalog No. FAB8626U100
Add to cart
This item is not returnable. View return policy
Description
Description
Protocadherin-19 Monoclonal antibody specifically detects Protocadherin-19 in Human samples. It is validated for Flow Cytometry.
Specifications
Specifications
Protocadherin-19
Monoclonal
0.2 mg/mL
Flow Cytometry 0.25-1 ug/10^6 cells
Q8TAB3
Mouse
IgG2b
RUO
Primary
Detects human Protocadherin-19 in direct ELISA and flow cytometry.
Flow Cytometry
921614
Alexa Fluor 350
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
PCDH19
HEK293 human embryonic kidney cell line transfected with human Protocadherin-19 Met1-Ser678 Accession # Q8TAB3
100 ug
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Monoclonal
Human
SDS
Documents
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For Research Use Only (RUO)