Learn More
Protocadherin Gamma A Mouse anti-Human, Mouse, Clone: S144-32, Invitrogen™
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen MA527649
Catalog No. PIMA527649
Add to cart
Description
Description
The protocadherin gamma gene cluster is one of three related clusters tandemly linked on chromosome five. These gene clusters have an immunoglobulin-like organization, suggesting that a novel mechanism may be involved in their regulation and expression. The gamma gene cluster includes 22 genes divided into 3 subfamilies. Subfamily A contains 12 genes, subfamily B contains 7 genes and 2 pseudogenes, and the more distantly related subfamily C contains 3 genes. The tandem array of 22 large, variable region exons are followed by a constant region, containing 3 exons shared by all genes in the cluster. Each variable region exon encodes the extracellular region, which includes 6 cadherin ectodomains and a transmembrane region. The constant region exons encode the common cytoplasmic region. These neural cadherin-like cell adhesion proteins most likely play a critical role in the establishment and function of specific cell-cell connections in the brain. Alternative splicing has been described for the gamma cluster genes.
Specifications
Specifications
Protocadherin Gamma A
Monoclonal
1 mg/ml
PBS with 50% glycerol and 0.1% sodium azide; pH 7.4
Q91Y11, Q9Y5H5
PCDH gamma A, PCDH-gamma-A, PCDHGA, Protocadherin gamma A, Protocadherin gamma subfamily A, Gamma Protocadherin A (pan), Pan-Gamma-Protocadherin-A, Pan Gamma Protocadherin A
Mouse
IgG2b
100 μg
-20°C
Primary
192161, 9752
Immunocytochemistry, Immunofluorescence, Western Blot
S144-32
Unconjugated
PCDHA9
Liquid
PCDHA9
Fusion protein amino acids 720-804 (variable cytoplasmic domain) of mouse Gamma-protocadherin-A3. Human: 80% identity (68/84 amino acids identical) >75% identity with other Gamma-protocadherin-A proteins
Protein G
RUO
Antibody
Monoclonal
Human, Mouse