Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Protocadherin gamma C3 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
Antigen | Protocadherin gamma C3 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
Protocadherin gamma C3 Polyclonal specifically detects Protocadherin gamma C3 in Human samples. It is validated for Western Blot.Specifications
Protocadherin gamma C3 | |
Polyclonal | |
Rabbit | |
Q9UN70 | |
5098 | |
Synthetic peptides corresponding to PCDHGC3(protocadherin gamma subfamily C, 3) The peptide sequence was selected from the C terminal of PCDHGC3. Peptide sequence IKDNGEPSLSTTATLTVSVTEDSPEARAEFPSGSAPREQKKNLTFYLLLS. | |
Primary |
Western Blot | |
Unconjugated | |
RUO | |
cadherin-like 2, PC43, PC-43, PCDH2, PCDH-GAMMA-C3, protocadherin 2, protocadherin 43, protocadherin gamma subfamily C, 3, protocadherin gamma-C3, protocadherin-2, Protocadherin-43 | |
PCDHGC3 | |
IgG |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title