Learn More
Protocadherin gamma C3 Mouse anti-Human, Mouse, Biotin, Clone: S174B-27, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP242203B
Catalog No. NBP242203B
Add to cart
Description
Description
Protocadherin gamma C3 Monoclonal antibody specifically detects Protocadherin gamma C3 in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry, Immunofluorescence.
Specifications
Specifications
Protocadherin gamma C3
S174B-27
Western Blot, Immunocytochemistry/Immunofluorescence
Purified
PCDHGC3
Fusion protein amino acids 720-804 (variable cytoplasmic domain) of mouse Gamma- protocadherin-C3. Human: 97% identity (83/85 amino acids identical). Rat: 97% identity (83/85 amino acids identical). No significant identity with other Gamma-protocadherin proteins.
Protein G purified
Store at 4C in the dark.
Monoclonal
No significant identity with other Gamma-protocadherin proteins.Detects approx 100kDa.
Western Blot, Immunocytochemistry, Immunofluorescence
Biotin
PBS with 0.05% Sodium Azide
cadherin-like 2, PC43, PC-43, PCDH2, PCDH-GAMMA-C3, protocadherin 2, protocadherin 43, protocadherin gamma subfamily C, 3, protocadherin gamma-C3, protocadherin-2, Protocadherin-43
Mouse
IgG1
0.1 ml
Primary
5098
Human, Mouse