Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PROTOCOL™ 10% Buffered Formalin, Fisher HealthCare™
Click to view available options
Quantity:
32 oz.
1 gal.
2.5 gal.
5 gal.
55 gal.
Packaging:
Cubitainer
Metal Drum
Poly Bottle
Specifications
Packaging | Poly Bottle |
Quantity | 1 gal. |
Chemical Name or Material | Fisher PROTOCOL™ 10% Buffered Formalin |
Safety and Handling
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction