Learn More
Fisher Healthcare™ PROTOCOL™ Parasitology Ancillary Products, Trichrome Stain
Standardize procedures for the routine collection, safe transportation, preservation and examination of stool specimens for intestinal parasites and enteric pathogens.
$80.90 - $287.00
Catalog Number | Volume (English) | Volume (Metric) | Description | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Volume (English) | Volume (Metric) | Description | Price | Quantity & Availability | ||||
23-005-38
|
16.91 oz. | 500 mL | TrichromeStain provides excellent contrast facilitating the detection of intestinal parasites while differentiating organisms from fecal debris and artifacts. |
Each for $287.00
|
|
||||
23-005-68
|
7.61 oz. | 225 mL | Ethyl Acetate |
Each for $94.50
|
|
||||
23-005-40
|
16.91 oz. | 500 mL | Zn-PVA Bulk - A mercury-free fixative for use as a preservative/fixative for protozoanm trophozoites. |
Each for $80.90
|
|
||||
Description
Standardize procedures for the routine collection, safe transportation, preservation and examination of stool specimens for intestinal parasites and enteric pathogens with Thermo Scientific™ PROTOCOL™ Parasitology Ancillary Products.
Trichrome Stain:
- Uses a rapid staining procedure to produce uniformly stained smears of intestinal protozoa, yeast and human cells
- Provides excellent contrast, facilitating the detection of intestinal parasites while differentiating organisms from fecal debris and artifacts
- Supplied in a 500mL bottle
Zinc Sulfite-Polyvinyl Alcohol (Zn-PVA) Bulk:
- Mercury-free fixative for protozoan trophozoites in a 500mL bottle
Ethyl Acetate:
- Supplied in a 225mL bottle
Note:
All PROTOCOL™ Parasitology products are mercury free.
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.