Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Healthcare™ PROTOCOL™ Parasitology Concentrate System

Catalog No. p-4534183 Shop All Fisherbrand Products
Click to view available options
Volume (English):
0.51 oz.
1.69 oz.
Volume (Metric):
15 mL
50 mL
This item is not returnable. View return policy
This item is not returnable. View return policy

Standardize procedures for the routine collection, safe transportation, preservation and examination of stool specimens for intestinal parasites and enteric pathogens.

Standardize procedures for the routine collection, safe transportation, preservation and examination of stool specimens for intestinal parasites and enteric pathogens with Thermo Scientific™ PROTOCOL™ Parasitology Concentrate Systems.

50mL Concentrate System

  • Complete system for concentrating and recovering helminth eggs, larvae and protozoan cysts from stool specimens
  • The concentration system creates a completely closed system that minimizes exposure to infectious agents and fits with any of the PROTOCOL System vials
  • Each 50mL Concentrate System Kit contains: 120 Filtering Devices, 120 Conical Centrifuge Tubes, 120 Screw Caps and 30mL Surfactant
  • Specimens collected and transported in 10% formalin, SAF or Clean vials may be used with this concentration system

15mL Concentrate System

  • Complete system for concentrating and recovering helminth eggs, larvae and protozoan cysts from stool specimens
  • Each 15mL Concentrate System Kit contains: 50 Filtering Devices, 50 Disposable 15mL centrifuge tubes and caps, Reagent A (Surfactant) and Reagent B (Ethyl Acetate)
  • Specimens collected and transported in 10% formalin, SAF or Clean vials may be used with this concentration system

Note:

All PROTOCOL Parasitology products are mercury free.

TRUSTED_SUSTAINABILITY

Specifications

Product Type Concentrate System
Description Concentrate System - A complete system for concentrating and recovering helminth eggs, larvae and protozoan cysts from stool specimens.
Includes 50 Filtering Devices, 50 Disposable 15mL Centrifuge Tubes and Caps, Reagent A (Surfactant), Reagent B (Ethyl Acetate)
Quantity 50/Pk.
Volume (English) 0.51 oz.
Volume (Metric) 15 mL
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.