Learn More
Thermo Scientific™ PROTOCOL™ Parasitology Three-Vial Kits
Standardize procedures for the routine collection, safe transportation, preservation and examination of stool specimens for intestinal parasites and enteric pathogens.
 View more versions of this product
Catalog No. 23-005-26
  Check Availability
Add to cart
Specifications
Description
Standardize procedures for the routine collection, safe transportation, preservation and examination of stool specimens for intestinal parasites and enteric pathogens with Thermo Scientific™ PROTOCOL™ Parasitology Three-Vial Kits.
Zinc Sulfite-Polyvinyl Alcohol (Zn-PVA)
  • Mercury-free fixative for use as a preservative/fixative for protozoan trophozoites
  • Slides fixed with Zn-PVA may be stained using trichrome or hematoxylin
Sodium Acetate Acetic Acid Formalin (SAF)
  • Mercury-free preservative/fixative ideally suited for use with helminth eggs and larvae, protozoan trophozoites and cysts, coccidian oocysts and microsporidian spores
  • May be stained using trichrome or hematoxylin
10% Buffered Formalin
  • All-purpose fixative appropriate for helminth eggs and larvae, and for protozoan cysts
  • Buffering system helps to maintain organism morphology
  • Fixed specimens may be examined directly or concentrated for recovery of eggs, larvae and protozoan cysts
Modified Cupric Sulfate-Polyvinyl Alcohol (Modified Cu-PVA)
  • Acts as a preservative/fixative for protozoan trophozoites
  • Slides fixed with Modified PVA may be stained using trichrome or hematoxylin
Culture and Sensitivity (C & S Media)
  • For collecting and transporting enteric bacterial pathogens such as Yersinia, Salmonella, Shigella, Campylobacter, and CholeraVibrio
  • Bacterial population is maintained for optimum recovery, up to 96 hours
Clean Vials
  • Empty, 30mL vials for transporting specimens
Ordering Alerts:
Supplied as 10 packs of three vials each
Note:
All PROTOCOL Parasitology products are mercury free.
Description & Specifications
Specifications
SAF/Clean/C and S
10 Pk.
10 kits per pack
Accessories