Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PROX2 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548392

Catalog No. PA548392

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Transcription regulator. Does not seem to be essential for embryonic development and postnatal survival.
Specifications

Specifications

PROX2
Polyclonal
PBS with 0.09% sodium azide
Q3B8N5
Prospero homeobox protein 2, Homeobox prospero-like protein PROX2, PROX-2, PROX2
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
283571
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
PROX2
Liquid
PROX2
KLH conjugated synthetic peptide between 511-539 amino acids from the C-terminal region of human PROX2.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit