Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Proxyfan oxalate, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
50 mg
Description
High affinity histamine H3 receptor ligand (pKi = 8.62) that acts as a protean agonist at recombinant and native receptors. In functional studies, activity ranges from full agonist to inverse agonist depending on system used. Displays partial agonist effects on cAMP inhibition and MAPK activity, neutral antagonist activity on histamine release and partial inverse agonism of [3H]-arachidonic acid release.
Specifications
Quantity | 50 mg |
Formula Weight | 306.32 |
Percent Purity | >98% |
Chemical Name or Material | Proxyfan oxalate |
Safety and Handling
Recommended Storage | Desiccate at Room Temperature |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction