Learn More
Invitrogen™ PRP19 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA593160
Description
Immunogen sequence: MSLICSISNE VPEHPCVSPV SNHVYERRLI EKYIAENGTD PINNQPLSEE QLIDIKVAHP IRPKPPSATS IPAILKALQD EWDAVMLHSF TLRQQLQTTR QELSHALYQH DAACRVIARL TKEVTAAREA LATLKPQAGL IVPQAVPSSQ PSVVGAGEPM DLGELVGMTP EIIQKLQDKA TVLTTERKKR GKTVPEELVK PEELSKYRQV ASHVGLHSAS; Positive Samples: SW620; Cellular Location: Cytoplasm, Lipid droplet, Nucleus, cytoskeleton, nucleoplasm, spindle.
PSO4 is the human homolog of yeast Pso4, a gene essential for cell survival and DNA repair.
Specifications
PRP19 | |
Polyclonal | |
Unconjugated | |
Prpf19 | |
AA617263; AL024362; D19Wsu55e; hPSO4; Neuronal differentiation-related gene protein; NMP200; Nuclear matrix protein 200; nuclear matrix protein NMP200 related to splicing factor PRP19; nuclear matrix protein SNEV; pre-mRNA processing factor 19; pre-mRNA-processing factor 19; PRP19; prp19 beta; PRP19/PSO4 homolog; PRP19/PSO4 pre-mRNA processing factor 19 homolog; PRP19/PSO4 pre-mRNA processing factor 19 homolog (S. cerevisiae); PRPF19; PSO4; psoralen 4; RING-type E3 ubiquitin transferase PRP19; Senescence evasion factor; SNEV; UBOX4 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
246216, 27339, 28000 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
1.5 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q99KP6, Q9JMJ4, Q9UMS4 | |
Prpf19 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-220 of human PRP19 (NP_0553171). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.