Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PRPF31 Polyclonal Antibody

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA528630

Catalog No. PIPA528630


Only null left
Add to Cart

Description

Description

Recommended positive controls: A549, HeLa, Jurkat, NT2D1. Predicted reactivity: Rhesus Monkey (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

This gene encodes a component of the spliceosome complex and is one of several retinitis pigmentosa-causing genes. When the gene product is added to the spliceosome complex, activation occurs.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

PRPF31
Polyclonal
Unconjugated
PRPF31
1500019O16Rik; 2810404O06Rik; AW554706; DKFZp566J153; hPrp 31; hPrp31; NY BR 99; NY-BR-99; pre-mRNA processing factor 31; pre-mRNA-processing factor 31; Protein 61K; PRP 31; Prp31; PRP31 pre-mRNA processing factor 31 homolog; PRP31 pre-mRNA processing factor 31 homolog (yeast); PRPF 31; Prpf31; RP 11; RP11; serologically defined breast cancer antigen NY-BR-99; SNRNP61; U4/U6 small nuclear ribonucleoprotein Prp31; U4/U6 snRNP 61 kDa protein
Rabbit
Antigen affinity chromatography
RUO
26121
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunoprecipitation, Western Blot
1 mg/mL
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
Q8WWY3
PRPF31
Synthetic peptide corresponding to a region within amino acids 437 and 499 of Human PRPF31.
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.