Learn More
PRPH Mouse anti-Human, Clone: OTI3F4, lyophilized, TrueMAB™ Non-distribution product as customer accommodation.
Mouse Monoclonal Antibody
Manufacturer:  OriGeneSupplier Diversity Partner CF806181
Catalog No. 50-166-7719
Add to cart
Description
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Peripherin is a 57kD type III intermediate filament that is a specific marker for peripheral neurons, including enteric ganglion cells. Peripherin is expressed in the developing peripheral nervous system and is highly enriched in neuronal derivatives of the neural crest. Peripherin offers an advantage over other neural markers, such as S100 and NSE in that it does not stain chrmaffin cell types.
Specifications
Specifications
PRPH
Monoclonal
Unconjugated
PRPH
Lyophilized
PRPH
Full length human recombit protein of human PRPH produced in HEK293T cell.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Immunohistochemistry (Paraffin)
OTI3F4
PBS with 8% trehalose and no preservative; pH 7.3
P41219
NEF4, PRPH1
Mouse
IgG1
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
5630