Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PRPK Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$149.00 - $329.00

Specifications

Antigen PRPK
Immunogen Synthetic peptides corresponding to TP53RK (TP53 regulating kinase) The peptide sequence was selected from the N terminal of TP53RK. Peptide sequence MAAARATTPADGEEPAPEAEALAAARERSSRFLSGLELVKQGAEARVFRG.
Host Species Rabbit
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP16916620 View Documents Novus BiologicalsSupplier Diversity Partner
NBP16916620UL
20ul Each for $149.00
Add to cart
 
NBP169166 View Documents Novus BiologicalsSupplier Diversity Partner
NBP169166
100 ul Each for $329.00
Add to cart
 
Description

Description

PRPK Polyclonal antibody specifically detects Antigen in Human samples. It is validated for Immunohistochemistry, Western Blotting.
Specifications

Specifications

PRPK
Rabbit
Polyclonal
Unconjugated
112858
Affinity Purified
Western Blot, Immunohistochemistry, Immunofluorescence
RUO
Synthetic peptides corresponding to TP53RK (TP53 regulating kinase) The peptide sequence was selected from the N terminal of TP53RK. Peptide sequence MAAARATTPADGEEPAPEAEALAAARERSSRFLSGLELVKQGAEARVFRG.
Primary
IgG
BUD32, C20orf64, chromosome 20 open reading frame 64, dJ101A2, dJ101A2.2, EC 2.7.11.1, Nori-2, Nori-2p, p53-related protein kinase, PRPK, TP53 regulating kinase, TP53-regulating kinase
PBS and 2% Sucrose with 0.09% Sodium Azide
Immunogen affinity purified
Store at -20C. Avoid freeze-thaw cycles.
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit