Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PRPS1 Mouse anti-Human, Alexa Fluor™ 750, Clone: 100000000000, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer: Novus Biologicals NBP242649AF750
Description
PRPS1 Monoclonal specifically detects PRPS1 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
PRPS1 | |
100000000000 | |
Western Blot, ELISA, Immunocytochemistry/Immunofluorescence | |
Purified, Conjugated | |
PRPS1 | |
Recombinant human PRPS1 (1-318aa) purified from E. coli | |
Protein A purified | |
Apoptosis, Core ESC Like Genes, Lipid and Metabolism, Stem Cell Markers | |
Primary | |
5631.0 |
Western Blot, ELISA, Immunocytochemistry, Immunofluorescence | |
Alexa Fluor 750 | |
50 mM sodium borate with 0.05% sodium azide | |
CMTX5ARTS, deafness, X-linked 2, perceptive, congenital, DFN2, DFNX1KIAA0967, dJ1070B1.2 (phosphoribosyl pyrophosphate synthetase 1), EC 2.7.6.1, Phosphoribosyl pyrophosphate synthase I, phosphoribosyl pyrophosphate synthetase 1, PPRibP, PRSI, PRS-I, ribose-phosphate diphosphokinase 1, ribose-phosphate pyrophosphokinase 1 | |
Mouse | |
IgG2a Kappa | |
100 μL | |
Store at 4°C in the dark. | |
Monoclonal | |
Human |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title