Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PRPS1 Polyclonal Antibody

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA527734

Catalog No. PIPA527734


Only null left
Add to Cart

Description

Description

Recommended positive controls: 293T, H1299, HeLa, HepG2, Molt-4, Raji. Predicted reactivity: Mouse (100%), Rat (100%), Xenopus laevis (95%), Rhesus Monkey (100%), Bovine (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

This gene encodes an enzyme that catalyzes the phosphoribosylation of ribose 5-phosphate to 5-phosphoribosyl-1-pyrophosphate, which is necessary for purine metabolism and nucleotide biosynthesis. Defects in this gene are a cause of phosphoribosylpyrophosphate synthetase superactivity, Charcot-Marie-Tooth disease X-linked recessive type 5 and Arts Syndrome.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

PRPS1
Polyclonal
Unconjugated
Prps1
2310010D17Rik; ARTS; C76571; C76678; CMTX5; deafness 2, perceptive, congenital; deafness, X-linked 2, perceptive, congenital; DFN2; DFNX1; dJ1070B1.2 (phosphoribosyl pyrophosphate synthetase 1); phosphoribosyl pyrophosphate synthase I; phosphoribosyl pyrophosphate synthetase 1; phosphoribosyl pyrophosphate synthetase I; PPRibP; PRPS1; Prps-1; PRSI; PRS-I; ribose-phosphate diphosphokinase 1; ribose-phosphate pyrophosphokinase 1
Rabbit
Antigen affinity chromatography
RUO
5631
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
1 mg/mL
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
P60891
Prps1
Recombinant protein fragment corresponding to a region within amino acids 142 and 307 of Human PRPS1.
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.