Learn More
PRPS1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 155491AP
Catalog No. 50-173-0307
Add to cart
Description
Description
This gene encodes an enzyme that catalyzes the phosphoribosylation of ribose 5-phosphate to 5-phosphoribosyl-1-pyrophosphate, which is necessary for purine metabolism and nucleotide biosynthesis. Defects in this gene are a cause of phosphoribosylpyrophosphate synthetase superactivity, Charcot-Marie-Tooth disease X-linked recessive type 5 and Arts Syndrome.
Specifications
Specifications
PRPS1
0.23 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P60891, P60892, Q9D7G0
ARTS, CMTX5, KIAA0967, PPRibP, PRPS1, PRS I, PRSI
Rabbit
IgG
150 μL
-20°C
Primary
19139, 29562, 5631
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Unconjugated
PRPS1
Liquid
Prps1
PRPS1 Fusion Protein Ag7907
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat