Learn More
Aviva Systems Biology Corp PRPSAP2 Antibody - N-terminal region (ARP56445_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP56445_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PRPSAP2. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N-terminal region of Human PRPSAP2. Alternate names: PAP41ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-61863