Learn More
PRPSAP2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 178141AP
Catalog No. 50-173-1810
Add to cart
Description
Description
The enzyme phosphoribosylpyrophosphate synthetase (PRS) catalyzes the formation of phosphoribosylpyrophosphate which is a substrate for synthesis of purine and pyrimidine nucleotides, histidine, tryptophan and NAD. PRS exists as a complex with two catalytic subunits and two associated subunits. This gene encodes a non-catalytic associated subunit of PRS.
Specifications
Specifications
PRPSAP2
0.19 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
O08618, O60256, Q8R574
PAP41, PRPSAP2
Rabbit
IgG
150 μL
-20°C
Primary
117272, 212627, 5636
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
PRPSAP2
Liquid
PRPSAP2
PRPSAP2 Fusion Protein Ag12132
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat