Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PRPSAP2 Mouse anti-Human, Clone: OTI1E3, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA525209

Catalog No. PIMA525209


Add to cart

Description

Description

The enzyme phosphoribosylpyrophosphate synthetase (PRS) catalyzes the formation of phosphoribosylpyrophosphate which is a substrate for synthesis of purine and pyrimidine nucleotides, histidine, tryptophan and NAD. PRS exists as a complex with two catalytic subunits and two associated subunits. This gene encodes a non-catalytic associated subunit of PRS.
Specifications

Specifications

PRPSAP2
Monoclonal
0.7 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
O60256
PAP41, 41 kDa phosphoribosypyrophosphate synthetase-associated protein, PRPP synthase-associated protein 2, phosphoribosyl pyrophosphate synthase-associated protein 2
Mouse
IgG1
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
5636
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
OTI1E3
Unconjugated
PRPSAP2
Liquid
PRPSAP2
Full length human recombit protein of PRPSAP2 produced in HEK293T cell
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit