Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PRR15L Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA546599
Description
Peptide sequence: RLEKIVDKST KGKHVKVSNS GRFKEKKKVR ATLAENPNLF DDHEEGRSSK Sequence homology: Human: 100%.
The function of this protein remains unknown.
Specifications
PRR15L | |
Polyclonal | |
Unconjugated | |
PRR15L | |
ATAD4; ATPase family, AAA domain containing 4; proline rich 15 like; proline rich 15-like; proline-rich protein 15-like protein; Protein ATAD4; PRR15L; testicular tissue protein Li 157 | |
Rabbit | |
Affinity chromatography | |
RUO | |
79170 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q9BU68 | |
PRR15L | |
Synthetic peptide directed towards the C-terminal of human PRR15L (aa 54-103). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction