Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PRR16 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5116725
Description
Positive Samples: Mouse brain, Mouse kidney Immunogen sequence: PNPPPPPPRL TPVKCEDPKR VVPTANPVKT NGTLLRNGGL PGGPNKIPNG DICCIPNSNL DKAPVQLLMH RPEKDRCPQA GPRERVRFNE KVQYHGYCPD C.
Regulator of cell size that promotes cell size increase independently of mTOR and Hippo signaling pathways. Acts by stimulating the translation of specific mRNAs, including those encoding proteins affecting mitochondrial functions. Increases mitochondrial mass and respiration.
Specifications
PRR16 | |
Polyclonal | |
Unconjugated | |
PRR16 | |
5430406M13Rik; AI607429; DSC54; LARGEN; mesenchymal stem cell protein DSC54; proline rich 16; proline-rich protein 16; Protein Largen; prr16; RGD1564528 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
71373 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
0.61 mg/mL | |
PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
A3KMN5 | |
PRR16 | |
A synthetic peptide corresponding to a sequence within amino acids 50-150 of human PRR16 (NP_0012950161). | |
100 μL | |
Primary | |
Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction