Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PRR18 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
$436.00
Specifications
Antigen | PRR18 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP170683
|
Novus Biologicals
NBP170683 |
100 μL |
Each of 1 for $436.00
|
|
Description
PRR18 Polyclonal specifically detects PRR18 in Human samples. It is validated for Western Blot.Specifications
PRR18 | |
Polyclonal | |
Rabbit | |
Human | |
proline rich 18 | |
PRR18 | |
IgG | |
Affinity Purified |
Western Blot | |
Unconjugated | |
RUO | |
PBS and 2% Sucrose with 0.09% Sodium Azide | |
285800 | |
Synthetic peptides corresponding to PRR18(proline rich region 18) The peptide sequence was selected from the middle region of PRR18. Peptide sequence LPARAAGPRRGGPASDPDAPPTAGQGRRAPPPGAQLLHGGLQVPQLSPRP. | |
Primary | |
Store at -20C. Avoid freeze-thaw cycles. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title