Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PRR19 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548290

Catalog No. PA548290

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

PRR19
Polyclonal
PBS with 0.09% sodium azide
A6NJB7
Proline-rich protein 19, PRR19
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
284338
Flow Cytometry, Western Blot
Unconjugated
PRR19
Liquid
PRR19
KLH conjugated synthetic peptide between 208-236 amino acids from the Central region of human PRR19.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit