Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PRR5L Antibody, DyLight 350, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP282025UV
This item is not returnable.
View return policy
Description
PRR5L Polyclonal antibody specifically detects PRR5L in Human, Mouse samples. It is validated for Western Blot, ELISA, ImmunofluorescenceSpecifications
PRR5L | |
Polyclonal | |
50mM Sodium Borate | |
Rabbit | |
Peptide affinity purified | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Western Blot, ELISA, Immunofluorescence | |
DyLight 350 | |
FLJ14213MGC16218, FLJ22630, proline rich 5 like, Protein observed with Rictor-2, protor-2, PROTOR2proline-rich protein 5-like | |
PRR5L antibody was raised against a 17 amino acid synthetic peptide near the center of human PRR5L. The immunogen is located within amino acids 220 - 270 of PRR5L. Amino Acid Squence: SGPTYTLARRHSRVRPKV | |
0.1 mL | |
Cancer, Cell Cycle and Replication | |
79899 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Product Content Correction