Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PRR7 Mouse anti-Human, Mouse, Rat, DyLight™ 650, Clone: TRAP3/10, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP130424C

Catalog No. NBP130424C


Add to cart

Description

Description

PRR7 Monoclonal antibody specifically detects PRR7 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry, Immunofluorescence.
Specifications

Specifications

PRR7
TRAP3/10
Western Blot, Immunocytochemistry/Immunofluorescence
Conjugated, Purified
PRR7
Recombinant C-terminal half of the intracellular domain of human PRR7/TRAP3 (amino acids 126-253)
Protein A purified
Store at 4°C in the dark.
Monoclonal
The mouse monoclonal antibody TRAP3/10 recognizes an epitope located in the C-terminal part of the intracellular domain of PRR7/TRAP3 (amino acids 126-253 of human PRR7/ TRAP3), a 28 kDa proline-rich membrane protein presumably associated with NMDA receptor complex.
Immunocytochemistry, Immunofluorescence, Western Blot
DyLight™ 650
50 mM sodium borate with 0.05% sodium azide
MGC10772, proline rich 7 (synaptic), proline-rich protein 7, Synaptic proline-rich membrane protein
Mouse
IgG2a
0.1 mL
Primary
80758.0
Human, Mouse, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit