Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PRRS Virus GP3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA5119591

This item is not returnable.
View return policy
Description
Store as concentrated solution. Centrifuge briefly prior to opening vial. For short-term storage (1-2 weeks), store at 4°C. For long-term storage, aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
PRRS Virus GP3 Polyclonal antibody specifically detects PRRS Virus GP3 in Virus samples. It is validated for Western BlotSpecifications
PRRS Virus GP3 | |
Polyclonal | |
Unconjugated | |
Porcine reproductive and respiratory syndrome virus ORF3; PRRSV ORF3 | |
Recombinant protein encompassing a sequence within the center region of PRRS virus GP3 protein (PRRS virus (strain PrimePac PRRS vaccine). The exact sequence is proprietary. | |
100 μL | |
Primary | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
1.3 mg/mL | |
PBS with 20% glycerol and 0.025% Proclin 300; pH 7 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
Virus | |
Antibody | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title