Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PRRSV Nucleocapsid Rabbit anti-Virus, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP313447
This item is not returnable.
View return policy
Description
PRRSV Nucleocapsid Polyclonal antibody specifically detects PRRSV Nucleocapsid in Virus samples. It is validated for Western Blot, ImmunofluorescenceSpecifications
PRRSV Nucleocapsid | |
Polyclonal | |
PBS, 20% Glycerol (pH7) | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
Virus | |
Purified |
Western Blot, Immunofluorescence | |
Unconjugated | |
Nucleocapsid protein, Nucleoprotein, Porcine reproductive and respiratory syndrome virus N protein, Protein N | |
Full length PRRS Virus Nucleocapsid recombinant protein. (strain Lelystad) | |
100 μg | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title