Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PRRT1 Mouse anti-Human,Mouse,Rat, Unconjugated, Clone: L102/45, MilliporeSigma™

Mouse Monoclonal Antibody

$386.00 - $391.00

Specifications

Antigen PRRT1
Applications Western Blot
Conjugate Unconjugated
Format Purified
Host Species Mouse
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MABN18671MI
View Documents
MilliporeSigma
MABN1867100UL
100 μL
Each for $386.00
Add to cart
 
MABN18672MI
View Documents
MilliporeSigma
MABN186725UL
25 μL
Each for $391.00
Add to cart
 
Description

Description

PRRT1 Monoclonal antibody specifically detects PRRT1 in Human,Mouse,Rat samples. It is validated for Western Blotting.
Specifications

Specifications

PRRT1
Unconjugated
Mouse
Proline-rich transmembrane protein 1;Dispanin subfamily D member 1;DSPD1;Synapse differentiation-induced protein 4
GST-tagged recombinant fragment corresponding to the first 66 amino acids from the N-terminal region of rat Proline-rich transmembrane protein 1 (PRRT1).
Primary
Stable for 1 year at 2 to 8°C from date of receipt.
Western Blot
Purified
Human, Mouse, Rat
Prrt1, Ng5
Monoclonal
Protein G Purified
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit