Learn More
Aviva Systems Biology Corp Prrxl1 Antibody - C-terminal region (ARP37845_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP37845_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Prrxl1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the c terminal region of mouse Prrxl1. Alternate names: Drg11, DrgxApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-62883