Learn More
PRSS2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 150051AP
Catalog No. 50-172-9903
Add to cart
Description
Description
This gene belongs to the trypsin family of serine proteases and encodes anionic trypsinogen. It is part of a cluster of trypsinogen genes that are located within the T cell receptor beta locus. Enzymes of this family cleave peptide bonds that follow lysine or arginine residues. This protein is found at high levels in pancreatic juice and its upregulation is a characteristic feature of pancreatitis. This protein has also been found to activate pro-urokinase in ovarian tumors, suggesting a function in tumor invasion. In addition, this enzyme is able to cleave across the type II collagen triple helix in rheumatoid arthritis synovitis tissue, potentially participating in the degradation of type II collagen-rich cartilage matrix. Alternative splicing results in multiple transcript variants.
Specifications
Specifications
PRSS2
0.27 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P00763, P07146, P07478
Anionic trypsinogen, PRSS2, Serine protease 2, TRY2, TRY8, TRYP2, Trypsin 2, Trypsin II
Rabbit
IgG
150 μL
-20°C
Primary
22072, 25052, 5645
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
PRSS2
Liquid
PRSS2
PRSS2 Fusion Protein Ag6461
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat