Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.

PRSS21 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA549493

Catalog No. PA549493

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Could regulate proteolytic events associated with testicular germ cell maturation.
Specifications

Specifications

PRSS21
Polyclonal
PBS with 0.09% sodium azide
Q9Y6M0
Testisin, 3421-, Eosinophil serine protease 1, ESP-1, Serine protease 21, PRSS21, ESP1, TEST1
Rabbit
IgG
200 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
10942
Western Blot
Unconjugated
PRSS21
Liquid
PRSS21
KLH conjugated synthetic peptide between 20-54 amino acids from human PRSS21.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit