Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PRSS21 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA5107225
Description
Sequence homology: Guinea Pig: 83%; Human: 100%; Mouse: 92%; Rabbit: 92%; Rat: 92%
PRSS21 Polyclonal antibody specifically detects PRSS21 in Human samples. It is validated for Western Blot.Specifications
PRSS21 | |
Polyclonal | |
Unconjugated | |
Prss21 | |
1700023E12Rik; EC 3.4.21; eosinophil serine protease 1; eosinophil serine protease-1; ESP1; ESP-1; mT4; protease, serine 21; protease, serine, 21; protease, serine, 21 (testisin); PRSS21; Resp-1; Serine protease 21; serine protease from eosinophils; TESP5; TEST1; TEST1testisin; testicular serine protease 5; testisin; Tryptase 4; UNQ266/PRO303 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
10942 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide; pH 7.4 | |
Q9Y6M0 | |
Prss21 | |
Synthetic peptide located within the following region: RRVITSRIVG GEDAELGRWP WQGSLRLWDS HVCGVSLLSH RWALTAAHCF | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction