Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PRSS33 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA563521
Description
Immunogen sequence: RALPAEYRVR LGALRLGSTS PRTLSVPVRR VLLPPDYSED GARGDLALLQ LR Highest antigen sequence identity to the following orthologs: Mouse - 71%, Rat - 73%.
Serine protease that has amidolytic activity, cleaving its substrates before Arg residues.
Specifications
PRSS33 | |
Polyclonal | |
Unconjugated | |
Prss33 | |
Eos; mT6; protease, serine 33; protease, serine, 33; Prss33; Serine protease 33; serine protease EOS; tryptase 6; Tryptase-6 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
260429 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8NF86 | |
Prss33 | |
Recombinant protein corresponding to Human PRSS33. Recombinant protein control fragment (Product #RP-98364). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction