Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PRSS33 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA550126

Catalog No. PA550126


Add to cart

Description

Description

The antibody detects endogenous levels of total PRSS33 protein.

Serine protease that has amidolytic activity, cleaving its substrates before Arg residues.
Specifications

Specifications

PRSS33
Polyclonal
Unconjugated
PRSS33
Liquid
PRSS33
Synthesized peptide derived from internal of human PRSS33.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Western Blot
1 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Q8NF86, Q80WM7
Serine protease 33 [Precursor], EC 3.4.21.-, Serine protease EOS, PRSS33,
Rabbit
IgG
100 μL
-20°C
Primary
260429, 353130
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit