Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PRSS33 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA549492

Catalog No. PA549492

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Serine protease that has amidolytic activity, cleaving its substrates before Arg residues.
Specifications

Specifications

PRSS33
Polyclonal
PBS with 0.09% sodium azide
Q8NF86
Serine protease 33, 3421-, Serine protease EOS, PRSS33
Rabbit
IgG
200 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
260429
Western Blot
Unconjugated
PRSS33
Liquid
PRSS33
KLH conjugated synthetic peptide between 105-128 amino acids from the Central region of human PRSS33.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit