Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PRSS36 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA549250

Catalog No. PA549250

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Serine protease. Hydrolyzes the peptides N-t-Boc-Gln- Ala-Arg-AMC and N-t-Boc-Gln-Gly-Arg-AMC and, to a lesser extent, N-t-Boc-Ala-Phe-Lys-AMC and N-t-Boc-Val-Leu-Lys-AMC. Has a preference for substrates with an Arg instead of a Lys residue in position P1.
Specifications

Specifications

PRSS36
Polyclonal
PBS with 0.09% sodium azide
Q5K4E3
Polyserase-2, 3421-, Polyserine protease 2, Serine protease 36, PRSS36
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
146547
Western Blot
Unconjugated
PRSS36
Liquid
PRSS36
KLH conjugated synthetic peptide between 288-317 amino acids from the Central region of human PRSS36.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit