Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PRSS42 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA549483

Catalog No. PA549483

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

PRSS42
Polyclonal
PBS with 0.09% sodium azide
Q7Z5A4
Serine protease 42, 3421-, Testis serine protease 2, PRSS42, TESSP2
Rabbit
IgG
200 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
339906
Western Blot
Unconjugated
PRSS42
Liquid
PRSS42
KLH conjugated synthetic peptide between 207-241 amino acids from the Central region of human PRSS42.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit