Learn More
PRTN3 Mouse anti-Human, Clone: OTI5B8, lyophilized, TrueMAB™ Non-distribution product as customer accommodation.
Mouse Monoclonal Antibody
Manufacturer:  OriGeneSupplier Diversity Partner CF807274
Catalog No. 50-166-8186
Add to cart
Description
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Polymorphonuclear leukocyte serine protease that degrades elastin, fibronectin, laminin, vitronectin, and collagen types I, III, and IV (in vitro) and causes emphysema when administered by tracheal insufflation to hamsters.
Specifications
Specifications
PRTN3
Monoclonal
Unconjugated
PRTN3
Lyophilized
PRTN3
Human recombit protein fragment corresponding to amino acids 28-248 of human PRTN3 produced in E.coli.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Immunohistochemistry (Paraffin)
OTI5B8
PBS with 8% trehalose and no preservative; pH 7.3
P24158
ACPA, AGP7, C-ANCA, CANCA, MBN, MBT, NP-4, NP4, P29, PR-3, PR3
Mouse
IgG2b
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
5657