Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PRUNE Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
Antigen | PRUNE |
---|---|
Concentration | 0.13 mg/mL |
Applications | Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-2174
![]() ![]() |
Proteintech Group Inc
185371AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
Phosphodiesterase (PDE) that has higher activity toward cAMP than cGMP, as substrate. It plays a role in cell proliferation, is able to induce cell motility and acts as a negative regulator of NME1.Specifications
PRUNE | |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
Q6AYG3, Q86TP1, Q8BIW1 | |
229589, 310664, 58497 | |
PRUNE Fusion Protein Ag13275 | |
Primary | |
-20°C | |
PRUNE1 |
0.13 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
DRES 17, DRES17, hPrune, HTCD37, Protein prune homolog, PRUNE, prune homolog (Drosophila) | |
PRUNE1 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title