Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PRUNE2 Polyclonal Antibody
Goat Polyclonal Antibody
Supplier: Invitrogen™ PA518725
Description
This antibody is predicted to react with canine based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 32,000.
PRUNE2 gene, also named BMCC1, encodes protein prune homolog 2. PRUNE2 gene are overlapping with PCA3, one of the most prostate cancer specific markers. PRUNE2 may play an important role in regulating differentiation, survival and aggressiveness of the tumor cells. Besides its high level of expression seen in the nervous system (brain, cerebellum and spinal cord), it is also expressed at high levels in noneuroblastoma, rhabdomyosarcoma, melanoma and some osteosarcoma cell lines, whereas at only low levels in cancer cell lines of liver, breast, thyroid and colon. The Calculated molecular weight of this protein is 340 kDa.
Specifications
PRUNE2 | |
Polyclonal | |
Unconjugated | |
PRUNE2 | |
6330414G02Rik; A214N16.3; A230083H22Rik; A330102H22Rik; bA214N16.3; BCH motif-containing molecule at the carboxyl terminal region 1; Bmcc1; BNIP2 motif containing molecule at the carboxyl terminal region 1; BNIP2 motif-containing molecule at the C-terminal region 1; BNIPXL; C9orf65; DKFZp762K117; KIAA0367; mKIAA0367; neuronal protein; olfaxin; protein prune homolog 2; prune homolog 2; prune homolog 2 (Drosophila); PRUNE2; RGD1311350; RP11-214N16.3; RP11-58J3.2; truncated PRUNE2 | |
Goat | |
Ammonium sulfate precipitation | |
RUO | |
158471 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
Q8WUY3 | |
PRUNE2 | |
Synthetic peptide sequence (MESEKISEKQEE) corresponding to the internal amino acids of PRUNE2 (aa 179-191). | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction