Learn More
PRUNE2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc 114581AP150UL

Description
The calculated molecular weight of this protein is 340 kDa. Our antibody specifically recognizes the 140-150 kDa band which is an isoform or cleaved fragment of PRUNE2.
PRUNE2 gene, also named BMCC1, encodes protein prune homolog 2. PRUNE2 gene are overlapping with PCA3, one of the most prostate cancer specific markers. PRUNE2 may play an important role in regulating differentiation, survival and aggressiveness of the tumor cells. Besides its high level of expression seen in the nervous system (brain, cerebellum and spinal cord), it is also expressed at high levels in noneuroblastoma, rhabdomyosarcoma, melanoma and some osteosarcoma cell lines, whereas at only low levels in cancer cell lines of liver, breast, thyroid and colon. The Calculated molecular weight of this protein is 340 kDa.Specifications
PRUNE2 | |
Polyclonal | |
Unconjugated | |
PRUNE2 | |
A214N16.3, BMCC1, BNIPXL, C9orf65, KIAA0367, Protein prune homolog 2, prune homolog 2 (Drosophila), PRUNE2, RP11 58J3.2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
158471, 293823, 353211 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.26 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q52KR3, Q5BJR4, Q8WUY3 | |
Prune2 | |
PRUNE2 Fusion Protein Ag2025 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.