PS 48, Tocris™

PDK1 activator

Manufacturer: R&D Systems 4087/10

Quantity 10mg

 View more versions of this product

Catalog No. 40-871-0

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    Assay Percent Range >98%
    Molecular Formula C17H15ClO2
    Formula Weight 286.75
    Synonym (2Z)-5-(4-Chlorophenyl)-3-phenyl-2-pentenoic acid
    Chemical Name or Material PS 48

    Specifications

    Formula Weight 286.75
    Quantity 10mg
    Assay Percent Range >98%
    Molecular Formula C17H15ClO2
    CAS 1180676-32-7
    Synonym (2Z)-5-(4-Chlorophenyl)-3-phenyl-2-pentenoic acid
    Recommended Storage Store at +4°C
    Chemical Name or Material PS 48

    Store at +4°C

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit