Learn More
Invitrogen™ PSAP Recombinant Rabbit Monoclonal Antibody (003)
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA531214
Description
This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire. This antibody has specificity for Mouse PSAP/Prosaposin.
PSAP is a highly conserved glycoprotein which is a precursor for 4 cleavage products: saposins A, B, C, and D. Each domain of the precursor protein is approximately 80 amino acid residues long with nearly identical placement of cysteine residues and glycosylation sites. Saposins A-D localize primarily to the lysosomal compartment where they facilitate the catabolism of glycosphingolipids with short oligosaccharide groups. The precursor protein exists both as a secretory protein and as an integral membrane protein and has neurotrophic activities.
Specifications
PSAP | |
Recombinant Monoclonal | |
1 mg/mL | |
PBS with no preservative | |
Q61207 | |
Psap | |
Recombinant Mouse PSAP/Prosaposin Protein (Met1-Asn557). | |
100 μL | |
Primary | |
Mouse | |
Antibody | |
IgG |
ELISA | |
003 | |
Unconjugated | |
Psap | |
A1 activator; AI037048; Cerebroside sulfate activator; Co-beta-glucosidase; Component C; CSAct; Dispersin; GLBA; Glucosylceramidase activator; Proactivator polypeptide; prosaposin; prosaposin (sulfated glycoprotein, sphingolipid hydrolase activator); Protein A; Protein C; Psap; SAP1; SAP-1; SAP-2; Saposin-A; Saposin-B; Saposin-B-Val; Saposin-C; Saposin-D; Sgp1; SGP-1; SGP1A; snoRNA MBII-198; Sphingolipid activator protein 1; Sphingolipid activator protein 2; sphingolipid activator protein-1; sulfated glycoprotein 1; Sulfatide/GM1 activator | |
Rabbit | |
Protein A | |
RUO | |
19156 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.