Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.

PSAPL1 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548368

Catalog No. PA548368

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

May activate the lysosomal degradation of sphingolipids.
Specifications

Specifications

PSAPL1
Polyclonal
PBS with 0.09% sodium azide
Q6NUJ1
Proactivator polypeptide-like 1, Saposin A-like, Saposin B-Val-like, Saposin B-like, Saposin C-like, Saposin D-like, PSAPL1
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
768239
Flow Cytometry, Western Blot
Unconjugated
PSAPL1
Liquid
PSAPL1
KLH conjugated synthetic peptide between 130-158 amino acids from the N-terminal region of human PSAPL1.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit