Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PSB 0474, Tocris Bioscience™

Click to view available options
Quantity:
1 mg
This item is not returnable. View return policy
917567-60-3
C17H18N2Na2O13P2
566.259
LEICCBKKYICWFH-BWBFMJMBSA-L
3-(2-Oxo-2-phenylethyl)-uridine-5'-diphosphate disodium salt
56972191
disodium;[[(2R,3S,4R,5R)-5-(2,4-dioxo-3-phenacylpyrimidin-1-yl)-3,4-dihydroxyoxolan-2-yl]methoxy-hydroxyphosphoryl] phosphate
C1=CC=C(C=C1)C(=O)CN2C(=O)C=CN(C2=O)C3C(C(C(O3)COP(=O)(O)OP(=O)([O-])[O-])O)O.[Na+].[Na+]
This item is not returnable. View return policy
917567-60-3
C17H18N2Na2O13P2
566.259
LEICCBKKYICWFH-BWBFMJMBSA-L
3-(2-Oxo-2-phenylethyl)-uridine-5'-diphosphate disodium salt
56972191
disodium;[[(2R,3S,4R,5R)-5-(2,4-dioxo-3-phenacylpyrimidin-1-yl)-3,4-dihydroxyoxolan-2-yl]methoxy-hydroxyphosphoryl] phosphate
C1=CC=C(C=C1)C(=O)CN2C(=O)C=CN(C2=O)C3C(C(C(O3)COP(=O)(O)OP(=O)([O-])[O-])O)O.[Na+].[Na+]

Potent and selective P2Y6 agonist

Potent and selective P2Y6 receptor agonist (EC50 values are 70, > 1000 and > 10000 nM for P2Y6, P2Y2 and P2Y4 receptors respectively). Induces contractions of rat isolated intrapulmonary arteries in vitro.
Quantity 1 mg
Formula Weight 566.26
Percent Purity >98%
Chemical Name or Material PSB 0474

Recommended Storage : Store at -20°C

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit