Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PSB 06126, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
50 mg
Description
Nucleoside triphosphate diphosphohydrolase 3 (NTPDase 3) inhibitor. Reported to inhibit rat NTPDase 3 at low micromolar concentrations and display selectivity over NTPDase 1 and NTPDase 2.
Specifications
Quantity | 10 mg |
Formula Weight | 466.44 |
Percent Purity | >98% |
Chemical Name or Material | PSB 06126 |
Safety and Handling
Recommended Storage : Store at Room Temperature