Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PSB 11 hydrochloride, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
50 mg
Description
Potent and selective antagonist for the human adenosine A3 receptor, with low affinity for the rat A3 receptor (Ki values are 2.3 and > 10000 nM respectively). Displays > 1000-fold selectivity over human A1 and A2A receptors (Ki values are 4.1 and 3.3 μM respectively) and > 180-fold selectivity over rat A1, rat A2A and mouse A2B receptors. Acts as an inverse agonist in the [35S]GTPγS binding assay in hA3-CHO cells (IC50 = 36 nM).
Specifications
Quantity | 10 mg |
Formula Weight | 331.8 |
Percent Purity | >98% |
Chemical Name or Material | PSB 11 hydrochloride |
Safety and Handling
Recommended Storage | Store at +4°C |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction