Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris™ PSB CB5
Selective GPR18 antagonist
$266.67 - $1122.22
Specifications
Molecular Formula | C20H17ClN2O2S |
---|---|
Purity | 98% |
Target | Additional Cannabinoid-related Compounds |
Chemical Name or Material | (2 Z)-2 [[3-[(4-Chlorophenyl)methoxy]phenyl]methylene]-6,7-dihydro-5 H-imidazo[2,1-b][1,3 ]thiazin-3(2 H)-one |
CAS | 1627710-30-8 |
Description
Selective GPR18 antagonist (IC50 = 0.279 μM). Exhibits ∽36-fold selectivity versus CB1 and GPR55 and 14-fold selectivity versus CB2 receptors. Inhibits N-Arachidonylglycine (Cat. No. 1445) induced acrosomal exocytosis in human spermatozoa.Specifications
C20H17ClN2O2S | |
Additional Cannabinoid-related Compounds | |
1627710-30-8 |
98% | |
(2 Z)-2 [[3-[(4-Chlorophenyl)methoxy]phenyl]methylene]-6,7-dihydro-5 H-imidazo[2,1-b][1,3 ]thiazin-3(2 H)-one |
Safety and Handling
Recommended Storage : Store at -20°C
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title